Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam N-Oleoylethanolamine, PPAR-alpha Agonist, Solid, >98% Purity, 100 mg

Catalog No. 502706854
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6854 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6854 Supplier Abcam Supplier No. AB141141100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potent PPAR-α agonist with anti-ischemic and neuroprotective effects.

  • Potent PPAR-α agonist: EC50 = 120 nM
  • Purity: >98%
  • Molecular Weight: 325.5 Da
  • Biological Activity: Anandamide analog that binds to cannabinoid receptor GPR119; shows anti-ischemic and neuroprotective effects in vivo
  • Administration: Orally active
  • Composition: 100% N-(2-Hydroxyethyl)oleamide
  • Form: Solid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Buffer 100% N-(2-Hydroxyethyl)oleamide
Content And Storage Store at -20°C.
Form Solid
Target Molecule PPAR-α, GPR119
Formulation 100% N-(2-Hydroxyethyl)oleamide
Quantity 100 mg
For Use With (Application) PPAR-alpha agonist, cannabinoid receptor GPR119 binding
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.