Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals N(omega)-Boc-N(beta)-Fmoc-L-beta-homolysine, 95%

Catalog No. AAH5218903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5218903 1 g
AAH52189MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5218903 Supplier Thermo Scientific Chemicals Supplier No. H5218903
Add to Cart

CAS: 203854-47-1 | C27H34N2O6 | 482.58 g/mol

N(μ)-Boc-N(beta)-Fmoc-L-beta-homolysine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N(µ)-Boc-N(beta)-Fmoc-L-beta-homolysine is used as pharmaceutical intermediate.

Solubility
Sparingly soluble bin water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 203854-47-1
Molecular Formula C27H34N2O6
Molecular Weight (g/mol) 482.58
MDL Number MFCD01863054
InChI Key SDBUQLGECIYUDT-LGWFVXIRNA-N
Synonym fmoc-l-beta-homolysine boc, s-3-9h-fluoren-9-yl methoxy carbonyl amino-7-tert-butoxycarbonyl amino heptanoic acid, fmoc-beta-holys boc-oh, fmoc-beta-homolys boc-oh, fmoc-l-nw-boc-beta-homolysine, 3s-7-1,1-dimethylethoxy carbonyl amino-3-9h-fluoren-9-ylmethoxy carbonyl amino heptanoic acid, 3s-7-tert-butoxycarbonyl amino-3-9h-fluoren-9-ylmethoxy carbonyl amino heptanoic acid, fmoc-l-beta-homolysine, ambotzfaa6700, fmoc-b-holys boc-oh
PubChem CID 2761531
IUPAC Name (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-7-[(2-methylpropan-2-yl)oxycarbonylamino]heptanoic acid
SMILES CC(C)(C)OC(=O)NCCCC[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 1 g
Beilstein 8022293
Solubility Information Sparingly soluble bin water.
Formula Weight 482.58
Percent Purity 95%
Chemical Name or Material N(omega)-Boc-N(beta)-Fmoc-L-beta-homolysine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.