Learn More
Abcam N Omega-Nitro-L-Arginine Methyl Ester Hydrochloride, >98% Purity, Solid, 100 mg
Description
- Molecular Weight: 269.69 Da
- Purity: >98%
- Form: Solid
- Function: Cell-permeable NOS inhibitor
- Key Properties: More soluble analog of arginine
- Inhibition Profile: pIC50 values are 4.3, 5.7 and 5.6 at human iNOS, nNOS and eNOS, respectively
- Composition: 100% methyl (2S)-2-amino-5-(N'-nitrocarbamimidamido)pentanoate hydrochloride
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Solid |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Target Molecule | Nitric Oxide Synthase |
| Quantity | 100 mg |
| For Use With (Application) | NOS inhibition |
| Storage Requirements | Store at 4°C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.