Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam N-Palmitoyl Phosphatidylethanolamine, Endogenous Lipid Anorectic Hormone, 1 mg

Catalog No. 502705389
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5389 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5389 Supplier Abcam Supplier No. AB1452191MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Endogenous lipid with potential as a therapeutic target for obesity, with >98% purity.

  • Molecular Weight: 930.4 g/mol
  • Purity: >98%
  • CAS Number: 108321-22-8
  • Storage Temperature: Store at -20°C
  • Shipping Temperature: Ambient - Can Ship with Ice
  • Composition: 100% N-Palmitoyl phosphatidylethanolamine
  • Biological Activity: Endogenous lipid anorectic hormone that reduces food intake and body weight in rats and mice, acting via the CNS

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule N-Palmitoyl Phosphatidylethanolamine
Buffer 100% N-Palmitoyl phosphatidylethanolamine
Substrate Properties Lipid-based Substrate
Content And Storage Store at -20°C.
Shipping Condition Ambient
Formulation 100% N-Palmitoyl phosphatidylethanolamine
Quantity 1 mg
For Use With (Application) Obesity-treatment research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.