Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentadecane, EEMD-Millipore

Catalog No. M1096070005 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1096070005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1096070005 Supplier MilliporeSigma Supplier No. 1.09607.0005

missing translation for 'validMainframeMsg'

Hydrocarbons C9 – C19

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-62-9
Molecular Formula C15H32
Molecular Weight (g/mol) 212.42
MDL Number MFCD00008990
InChI Key YCOZIPAWZNQLMR-UHFFFAOYSA-N
PubChem CID 12391
ChEBI CHEBI:28897
IUPAC Name pentadecane
SMILES CCCCCCCCCCCCCCC

Specifications

Packaging Glass ampoule
Explosion Limit 0.45 to 6.5 %(V)
Vapor Pressure < - 0.2 hPa (20°C)
Viscosity <7mm2/sec., kinematic (40°C)
Density Density: 0.77g/cm3 (20°)
Assay Percent Range ≥99.5%
Linear Formula CH3(CH2)13CH3
Quantity 5 mL
Formula Weight 212.41
Chemical Name or Material n-Pentadecane
Recommended Storage Store without restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.