accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 629-62-9

IUPAC Name: pentadecane
Synonyms: pentadecane pentadekan N-pentadecane
Molecular Weight (g/mol): 212.42
Molecular Formula: C15H32
InChi Key: YCOZIPAWZNQLMR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (6)
  • (13)
  • (4)
Boiling Point 
  • (4)
  • (4)
  • (3)
  • (6)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (4)
  • (3)
  • (1)
  • (11)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Physical Form 
  • (7)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (4)
  • (1)
  • (6)
  • (4)
  • (1)
  • (4)
  • (7)
  • (6)
  • (13)
  • (4)
  • (4)
  • (4)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (11)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (7)
  • (4)
  • (4)
  • (1)