Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentane, HPLC Grade, 99% min

Catalog No. p-4408277
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA22907K2 1 L
AA22907M1 4 x 1 L
AA22907K7 4 L
AA22907M6 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA22907K2 Supplier Thermo Scientific Chemicals Supplier No. 022907K2
Only null left
Add to Cart

Maximum level of impurities: Evaporation residue 3ppm, H2O 0.01% | CAS: 109-66-0 | C5H12 | 72.15 g/mol

n-Pentane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point 130°C
Color Colorless
Appearance Colorless liquid
Density 0.626 g/cm3
Quantity 1 L
Flash Point -49°C (-57°F)
Ignition Point 260°C (500°F)
Physical Form Liquid
Refractive Index 1.358
Assay Percent Range 99% min.
Vapor Pressure 554 hPa (416mm Hg) at 20°C
Solubility Information Miscible with water.
Boiling Point 36°C
Formula Weight 72.15
Grade HPLC
Chemical Name or Material n-Pentane
Show More Show Less
Hazard Category H225-H304-H336
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P301+P310-P303+P361+P353-P304+P340-P312-P331-P370+P378q-P501c
DOTInformation Hazard Class: 3, Packing Group: II
EINECSNumber 203-692-4
RTECSNumber RZ9450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.