Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentane, 99+%, for spectroscopy

Catalog No. p-216678
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167875000 500 mL Glass Bottle
AC167870025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167875000 Supplier Thermo Scientific Chemicals Supplier No. 167875000
Only null left
Add to Cart

n-Pentane, >-99%, C5H12, CAS Number-109-66-0, tetraspot, n-pentane, amyl hydride, pentan, pentanen, pentani, pentan polish, skellysolve a, tetrafume, tetrakil, 2.5L, 0.009 max. at 240nm, 0.018 max. at 230nm, 0.046 max. at 220nm, 0.222 max. at 210nm, 0.699 max. at 200nm, 99+%, 01, 130, 36.0 deg.C | CAS: 109-66-0 | C5H12 | 72.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
Synonym n-pentane, pentan, skellysolve a, pentanen, pentani, amyl hydride, tetrafume, tetrakil, tetraspot, pentan polish
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Melting Point -130.0°C
Absorbance 0.009 max. at 240nm, 0.018 max. at 230nm, 0.046 max. at 220nm, 0.222 max. at 210nm, 0.699 max. at 200nm
CAS Min % 99.0
CAS Max % 100.0
Density 0.6260g/mL
Quantity 500 mL
Flash Point −49°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.358
Residue after Evaporation 0.001% max.
Assay Percent Range 99+%
Specific Gravity 0.626
Linear Formula CH3(CH2)3CH3
Viscosity 0.25 mPa.s (20°C)
Beilstein 01, 130
Water 0.01% max.
Merck Index 15, 7228
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and most other common organic, solvents
Boiling Point 36.0°C
Formula Weight 72.15
Percent Purity ≥99%
Grade Spectroscopy
Chemical Name or Material n-Pentane, 99+%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.
EINECSNumber 203-692-4
RTECSNumber RZ9450000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.