Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Pentane, EMD-Millipore

Catalog No. M1097190005 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1097190005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1097190005 Supplier MilliporeSigma Supplier No. 1.09719.0005

missing translation for 'validMainframeMsg'

Hydrocarbons C5

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-66-0
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009498
InChI Key OFBQJSOFQDEBGM-UHFFFAOYSA-N
PubChem CID 8003
ChEBI CHEBI:37830
IUPAC Name pentane
SMILES CCCCC

Specifications

Packaging Glass ampoule
Explosion Limit 1.4 to 8 %(V)
Ignition Point 285°C
Vapor Pressure 570 hPa (20°C)
Density Density: 0.63g/cm3 (20°)
Assay Percent Range ≥99.7%
Linear Formula CH3(CH2)3CH3
Quantity 5 mL
Formula Weight 72.15
Chemical Name or Material n-Pentane
Recommended Storage Store at 15° to 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.