Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenyl-p-phenylenediamine, 98%

Catalog No. AC221002500
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC221002500 250 g Glass bottle
AC221001000 100 g Glass bottle
AC221000010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC221002500 Supplier Thermo Scientific Chemicals Supplier No. 221002500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 101-54-2 | C12H12N2 | 184.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-54-2
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.24
MDL Number MFCD00007850
InChI Key ATGUVEKSASEFFO-UHFFFAOYSA-N
Synonym 4-aminodiphenylamine, n-phenyl-p-phenylenediamine, n1-phenylbenzene-1,4-diamine, p-anilinoaniline, azosalt r, p-semidine, p-aminodiphenylamine, luxan black r, semidine, n-phenyl-1,4-phenylenediamine
PubChem CID 7564
ChEBI CHEBI:59038
IUPAC Name 4-N-phenylbenzene-1,4-diamine
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 69.0°C to 75.0°C
Boiling Point 354.0°C
Flash Point >250°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5NHC6H4NH2
Quantity 250 g
Beilstein 13, 76
Solubility Information Solubility in water: 0.6g/L (20°C). Other solubilities: soluble in alcohol and acetone
Formula Weight 184.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Phenyl-p-phenylenediamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause an allergic skin reaction.
Harmful if swallowed.
Very toxic to aquatic life
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENT
EINECSNumber 202-951-9
RTECSNumber ST3150000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.