Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenyl-p-phenylenediamine Monohydrochloride 99%, Thermo Scientific™

Catalog No. AC417680100
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC417680100 10 g Glass bottle
AC417680250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC417680100 Supplier Thermo Scientific Chemicals Supplier No. 417680100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2198-59-6
Molecular Formula C12H13ClN2
Molecular Weight (g/mol) 220.7
InChI Key PNEVDCGZQWFIKV-UHFFFAOYSA-N
Synonym n1-phenylbenzene-1,4-diamine hydrochloride, 4-aminodiphenylamine hydrochloride, eucanine grey b, 1,4-benzenediamine, n-phenyl-, monohydrochloride, fourrine grey b, durafur grey bn, fouramine grey a, futramine grey b, furro grey b, nako grey gb
PubChem CID 75146
IUPAC Name 4-N-phenylbenzene-1,4-diamine;hydrochloride
SMILES C1=CC=C(C=C1)NC2=CC=C(C=C2)N.Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Blue-Gray
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 10 g
Formula Weight 220.7
Percent Purity ≥98.5%
Physical Form Powder
Chemical Name or Material N-Phenyl-p-phenylenediamine monohydrochloride, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-599-4
RTECSNumber ST3551000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.