Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylglycine Potassium Salt 98.0+%, TCI America™
SDP

Catalog No. P017925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P017925G 25 g
P0179500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P017925G Supplier TCI America Supplier No. P017925G

Chemical Identifiers

CAS 19525-59-8
Molecular Formula C8H8KNO2
Molecular Weight (g/mol) 189.255
MDL Number MFCD00058994
InChI Key JINONCVJYBHUCP-UHFFFAOYSA-M
PubChem CID 23672325
IUPAC Name potassium;2-anilinoacetate
SMILES C1=CC=C(C=C1)NCC(=O)[O-].[K+]

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 189.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylglycine Potassium Salt
EINECSNumber (3)-0150
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.