Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phenylmaleimide 98.0+%, TCI America™
SDP

Catalog No. P0900100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0900100G 100 g
P090025G 25 g
P0900500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P0900100G Supplier TCI America Supplier No. P0900100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 941-69-5
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.171
MDL Number MFCD00005502
InChI Key HIDBROSJWZYGSZ-UHFFFAOYSA-N
Synonym n-phenylmaleimide, maleanil, maleinanil, maleimidobenzene, 1h-pyrrole-2,5-dione, 1-phenyl, 1-phenyl-1h-pyrrole-2,5-dione, maleimide, n-phenyl, 1-phenyl-2,5-dihydro-1h-pyrrole-2,5-dione, unii-9u9kt462vw, n-fenylimid kyseliny maleinove
PubChem CID 13662
IUPAC Name 1-phenylpyrrole-2,5-dione
SMILES C1=CC=C(C=C1)N2C(=O)C=CC2=O

Specifications

Melting Point 90°C
Color Yellow
Boiling Point 181°C
Quantity 100 g
UN Number 2811
Formula Weight 173.17
Percent Purity ≥98.0% (GC)
Physical Form Flakes
Chemical Name or Material N-Phenylmaleimide
EINECSNumber (5)-5727
RTECSNumber ON5950000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.