Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(Phenylthio)phthalimide 98.0+%, TCI America™
SDP

Catalog No. P125425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P125425G 25 g
P12545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P125425G Supplier TCI America Supplier No. P125425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14204-27-4
Molecular Formula C14H9NO2S
Molecular Weight (g/mol) 255.29
MDL Number MFCD00192396
InChI Key NMHKBABHRKQHOL-UHFFFAOYSA-N
PubChem CID 5127161
IUPAC Name 2-(phenylsulfanyl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES O=C1N(SC2=CC=CC=C2)C(=O)C2=CC=CC=C12

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 25 g
Formula Weight 255.29
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(Phenylthio)phthalimide
EINECSNumber (5)-0090
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.