Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Phthaloyl-L-glutamic Anhydride 98.0+%, TCI America™
SDP

Catalog No. P06201G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06201G 1 g
P06205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P06201G Supplier TCI America Supplier No. P06201G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25830-77-7
Molecular Formula C13H9NO5
Molecular Weight (g/mol) 259.217
MDL Number MFCD00067324
InChI Key ICDLEMPZXFCQEB-VIFPVBQESA-N
PubChem CID 11807393
IUPAC Name 2-[(3S)-2,6-dioxooxan-3-yl]isoindole-1,3-dione
SMILES C1CC(=O)OC(=O)C1N2C(=O)C3=CC=CC=C3C2=O

Specifications

Melting Point 205°C
Color White
Quantity 1 g
Formula Weight 259.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Phthaloyl-L-glutamic Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.