Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Propyl 3,4,5-trihydroxybenzoate, 98%

Catalog No. p-7081016
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1087706 5 g
AAA1087722 100 g
AAA1087736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1087706 Supplier Thermo Scientific Chemicals Supplier No. A1087706
Only null left
Add to Cart

CAS: 121-79-9 | C10H12O5 | 212.201 g/mol

n-Propyl 3,4,5-trihydroxybenzoate is an anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein. Also used as an antioxidant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Propyl 3,4,5-trihydroxybenzoate is an anti-fade reagent in fluorescence microscopy to reduce photobleaching of fluorescent probes such as rhodamine and fluorescein. Also used as an antioxidant.

Solubility
Soluble in ethanol (50 mg/ml) and water (0.35 g/100 ml at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-79-9
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.201
MDL Number MFCD00002196
InChI Key ZTHYODDOHIVTJV-UHFFFAOYSA-N
Synonym propyl gallate, n-propyl gallate, progallin p, tenox pg, nipagallin p, gallic acid, propyl ester, gallic acid propyl ester, nipa 49, benzoic acid, 3,4,5-trihydroxy-, propyl ester, 3,4,5-trihydroxybenzene-1-propylcarboxylate
PubChem CID 4947
ChEBI CHEBI:10607
IUPAC Name propyl 3,4,5-trihydroxybenzoate
SMILES CCCOC(=O)C1=CC(=C(C(=C1)O)O)O

Specifications

Melting Point 146°C to 150°C
Quantity 5 g
Beilstein 1877976
Merck Index 147859
Solubility Information Soluble in ethanol (50mg/ml) and water (0.35g/100 ml at 25°C).
Formula Weight 212.2
Percent Purity 0.98
Chemical Name or Material n-Propyl 3,4,5-trihydroxybenzoate
Hazard Category H302-H317-H319
Hazard Statement GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c
EINECSNumber 204-498-2
RTECSNumber LW8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.