Learn More
Bon Opus Biosciences Recombinant 2019-nCoV N Protein (Truncated) -1mg
SDP
Supplier:  Bon Opus Biosciences DRA581MG
Encompass_Preferred
Recombinant 2019-nCoV NP (Truncated) is produced by our E.coli expression system with a 6His tag at the N-terminus. Expression system: E. coli. Accession# QHD43423.2. Formulation: Supplied as a 0.2 μM filtered solution of PBS, pH 7.4. Bon Opus Cat# DRA58
Catalog No. 50-208-0550
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.