Learn More
Cayman Chemical N- S- 4-NItrophenoxyphenox 10g
SDP
Supplier:  Cayman Chemical 3474810 G
Encompass_Preferred
A phthalide with diverse biological activities inhibits TPA and A23187-induced histamine release from HMC-1 cells at 10 UM induces the production of ROS ER stress and apoptosis in HCT116 cells at 100 UM prevents ethanol-induced gastric ulcers in rats at 10 mg/kg decreases aggregation of amyloid-B in brain and improves spatial and learning memory in the APP/PS1 transgenic mouse model of Alzheimers disease at 2 mg/kg
Catalog No. 50-291-2358
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.