Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl 4-[4-(Dimethylamino)phenylazo]benzoate 98.0+%, TCI America™
SDP

Catalog No. S08571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08571G 1 g
S0857200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08571G Supplier TCI America Supplier No. S08571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 146998-31-4
Molecular Formula C19H18N4O4
Molecular Weight (g/mol) 366.377
MDL Number MFCD00467415
InChI Key IBOVDNBDQHYNJI-UHFFFAOYSA-N
Synonym 2,5-dioxopyrrolidin-1-yl 4-4-dimethylamino phenyl diazenyl benzoate, dabcyl se dye, dabcyl acid, se, 4-4-dimethylamino phenyl azo benzoic acid, succinimidyl ester, 4-4-dimethylamino phenylazo benzoic acid n-succinimidyl ester, dabcyl-nhs, e-2,5-dioxopyrrolidin-1-yl 4-4-dimethylamino phenyl diazenyl benzoate, 2,5-pyrrolidinedione, 1-4-4-dimethylamino phenyl azo benzoyl oxy, dabcyl-se; 4-4-dimethylamino phenylazo benzoic acid n-succinimidyl ester, dabcyl se, solid
PubChem CID 4206604
ChEBI CHEBI:52005
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 4-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)C(=O)ON3C(=O)CCC3=O

Specifications

Melting Point 242°C
Quantity 1 g
Formula Weight 366.38
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 4-[4-(Dimethylamino)phenylazo]benzoate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.