Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl 4-Azido-2,3,5,6-tetrafluorobenzoate 97.0+%, TCI America™
SDP

Catalog No. S09521G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09521G 1 g
S0952200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S09521G Supplier TCI America Supplier No. S09521G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 126695-58-7
Molecular Formula C11H4F4N4O4
Molecular Weight (g/mol) 332.17
MDL Number MFCD00467437
InChI Key HCTLWQSYGIBOFM-UHFFFAOYSA-N
Synonym 4-Azido-2,3,5,6-tetrafluorobenzoic Acid N-Succinimidyl Ester, ATFB-SE
PubChem CID 2762579
IUPAC Name 2,5-dioxopyrrolidin-1-yl 4-azido-2,3,5,6-tetrafluorobenzoate
SMILES FC1=C(F)C(C(=O)ON2C(=O)CCC2=O)=C(F)C(F)=C1N=[N+]=[N-]

Specifications

Melting Point 113°C
Color White
Quantity 1 g
UN Number 1325
Formula Weight 332.17
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 4-Azido-2,3,5,6-tetrafluorobenzoate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.