Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl [(tert-Butoxycarbonyl)aminooxy]acetate 98.0+%, TCI America™
SDP

Catalog No. U00961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00961G 1 g
U0096200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U00961G Supplier TCI America Supplier No. U00961G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 80366-85-4
Molecular Formula C11H16N2O7
Molecular Weight (g/mol) 288.26
MDL Number MFCD18382162
InChI Key ZZBOTLREHORFCK-UHFFFAOYSA-N
Synonym [(tert-Butoxycarbonyl)aminooxy]acetic Acid N-Succinimidyl Ester, N-Succinimidyl (Boc-aminooxy)acetate, (Boc-aminooxy)acetic Acid N-Succinimidyl Ester
PubChem CID 12098210
IUPAC Name 2,5-dioxopyrrolidin-1-yl 2-({[(tert-butoxy)carbonyl]amino}oxy)acetate
SMILES CC(C)(C)OC(=O)NOCC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 114°C
Color White
Quantity 1 g
Formula Weight 288.26
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl [(tert-Butoxycarbonyl)aminooxy]acetate
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.