Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Sulfinyl-p-toluenesulfonamide 70.0+%, TCI America™
SDP

Catalog No. T14611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14611G 1 g
T14615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14611G Supplier TCI America Supplier No. T14611G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 4104-47-6
Molecular Formula C7H7NO3S2
Molecular Weight (g/mol) 217.257
MDL Number MFCD00082512
InChI Key VKTSIMMJOIPMGE-UHFFFAOYSA-N
Synonym N-Thionyl-p-toluenesulfonamide
PubChem CID 562840
IUPAC Name 4-methyl-N-sulfinylbenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N=S=O

Specifications

Quantity 1 g
Formula Weight 217.26
Percent Purity ≥70.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Sulfinyl-p-toluenesulfonamide
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.