Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-3-(2-pyridyl)-L-alanine 98.0+%, TCI America™
SDP

Catalog No. B42931G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42931G 1 g
B42935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42931G Supplier TCI America Supplier No. B42931G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 71239-85-5
Molecular Formula C13H18N2O4
Molecular Weight (g/mol) 266.297
MDL Number MFCD00191190
InChI Key KMODKKCXWFNEIK-JTQLQIEISA-N
Synonym boc-l-2-pyridylalanine, s-2-tert-butoxycarbonyl amino-3-pyridin-2-yl propanoic acid, boc-3-2-pyridyl-l-alanine, boc-l-3-2-pyridyl-alanine, boc-ala 2-pyridyl-oh, boc-2-pal-oh, boc-3-2-pyridyl-ala-oh, boc-2'-pyridyl-l-ala, n-boc-3-2-pyridyl-l-alanine, boc-l-2-pyridylala
PubChem CID 2734482
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-pyridin-2-ylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=N1)C(=O)O

Specifications

Melting Point 141°C
Color White-Yellow
Quantity 1 g
Formula Weight 266.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-3-(2-pyridyl)-L-alanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.