Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. B33031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33031G 1 g
B33035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33031G Supplier TCI America Supplier No. B33031G

Chemical Identifiers

CAS 57292-45-2
Molecular Formula C14H17FNO4
Molecular Weight (g/mol) 282.29
MDL Number MFCD00079669
InChI Key RCXSXRAUMLKRRL-LLVKDONJSA-M
Synonym boc-d-phe 4-f-oh, boc-d-4-fluorophe, boc-d-4-fluorophenylalanine, boc-4-fluoro-d-phenylalanine, boc-4-fluoro-d-phe, n-tert-butoxycarbonyl-4-fluoro-d-phenylalanine, boc-p-fluoro-d-phe-oh, boc-4-fluoro-d-phe-oh, 2r-2-tert-butoxycarbonyl amino-3-4-fluorophenyl propanoic acid, boc-p-fluoro-d-phenylalanine
PubChem CID 2734493
IUPAC Name (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-fluorophenyl)propanoate
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CC=C(F)C=C1)C([O-])=O

Specifications

Melting Point 87°C
Color White
Quantity 1 g
Formula Weight 283.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-fluoro-D-phenylalanine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.