Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-4-nitro-D-phenylalanine 98.0+%, TCI America™
SDP

Catalog No. B36681G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36681G 1 g
B36685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36681G Supplier TCI America Supplier No. B36681G

Chemical Identifiers

CAS 61280-75-9
Molecular Formula C14H18N2O6
Molecular Weight (g/mol) 310.31
MDL Number MFCD00069880
InChI Key XBQADBXCNQPHHY-UHFFFAOYNA-N
Synonym boc-4-nitro-d-phenylalanine, boc-d-phe 4-no2-oh, boc-d-4-nitrophe, boc-d-4-no2-phe-oh, r-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, n-tert-butoxycarbonyl-4-nitro-d-phenylalanine, boc-d-4-nitrophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-nitrophenyl propanoic acid, d-phenylalanine, n-1,1-dimethylethoxy carbonyl-4-nitro, 2r-2-2-methylpropan-2-yl oxycarbonylamino-3-4-nitrophenyl propanoic acid
PubChem CID 2755957
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-3-(4-nitrophenyl)propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 1 g
Formula Weight 310.31
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-nitro-D-phenylalanine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.