Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-D-proline Methyl Ester 98.0+%, TCI America™
SDP

Catalog No. B47871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47871G 1 g
B47875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47871G Supplier TCI America Supplier No. B47871G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 73323-65-6
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.276
MDL Number MFCD00190828
InChI Key WVDGSSCWFMSRHN-MRVPVSSYSA-N
Synonym boc-d-pro-ome, boc-d-proline methyl ester, r-1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl 2r-pyrrolidine-1,2-dicarboxylate, n-boc-d-proline methyl ester, r-1-boc-pyrrolidine-2-carboxylic acid methyl ester, n-tert-butoxycarbonyl-d-proline methyl ester, o1-tert-butyl o2-methyl 2r-pyrrolidine-1,2-dicarboxylate, boc-d-proline-methyl ester, pubchem16716
PubChem CID 1277667
IUPAC Name 1-O-tert-butyl 2-O-methyl (2R)-pyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)OC

Specifications

Color Yellow
Quantity 1 g
Formula Weight 229.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-proline Methyl Ester
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.