Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-D-threonine Methyl Ester 98.0+%, TCI America™
SDP

Catalog No. B55371G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55371G 1 g
B55375G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55371G Supplier TCI America Supplier No. B55371G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 96099-84-2
Molecular Formula C10H19NO5
Molecular Weight (g/mol) 233.264
InChI Key MZMWAPNVRMDIPS-NKWVEPMBSA-N
Synonym N-Boc-D-threonine Methyl Ester, Boc-D-Thr-OMe
PubChem CID 40501164
IUPAC Name methyl (2R,3S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoate
SMILES CC(C(C(=O)OC)NC(=O)OC(C)(C)C)O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 233.26
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-threonine Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.