Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-L-isoleucine Hemihydrate 99.0+%, TCI America™
SDP

Catalog No. B118625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B118625G 25 g
B11865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B118625G Supplier TCI America Supplier No. B118625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13139-16-7
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00038324 MFCD00237550
InChI Key QJCNLJWUIOIMMF-YUMQZZPRSA-N
Synonym boc-l-isoleucine, boc-ile-oh, n-boc-l-isoleucine, n-tert-butoxycarbonyl-l-isoleucine, boc-dl-ile-oh, boc-l-ile-oh, 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoic acid, l-isoleucine, n-1,1-dimethylethoxy carbonyl, boc-isoleucine, n-t-boc-l-isoleucine
PubChem CID 2724762
IUPAC Name (2S,3S)-2-{[(tert-butoxy)carbonyl]amino}-3-methylpentanoic acid
SMILES CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 60°C
Color White
Quantity 25 g
Formula Weight 231.29
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-isoleucine Hemihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.