Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-L-norvaline 98.0+%, TCI America™
SDP

Catalog No. B43201G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43201G 1 g
B43205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43201G Supplier TCI America Supplier No. B43201G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53308-95-5
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.27
MDL Number MFCD00037268
InChI Key INWOAUUPYIXDHN-ZETCQYMHSA-N
Synonym boc-l-norvaline, boc-nva-oh, n-boc-l-norvaline, boc-norvaline, 2s-2-tert-butoxycarbonyl amino pentanoic acid, boc-l-2-aminovaleric acid, n-1,1-dimethylethoxy carbonyl-l-methionine, n-tert-butoxycarbonyl-l-norvaline, s-2-tert-butoxycarbonyl amino pentanoic acid, 2s-2-tert-butoxy carbonylamino pentanoic acid
PubChem CID 2733748
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}pentanoic acid
SMILES CCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Melting Point 68°C
Color White
Quantity 1 g
Formula Weight 217.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-norvaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.