Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-L-prolinamide 98.0+%, TCI America™
SDP

Catalog No. B37561G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37561G 1 g
B37565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37561G Supplier TCI America Supplier No. B37561G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35150-07-3
Molecular Formula C10H18N2O3
Molecular Weight (g/mol) 214.265
MDL Number MFCD00190827
InChI Key PITJAAIPVBVRAO-ZETCQYMHSA-N
Synonym boc-pro-nh2, 1-boc-l-prolinamide, boc-l-proline amide, l-1-n-boc-prolidinamide, s-tert-butyl 2-carbamoylpyrrolidine-1-carboxylate, boc-l-prolinamide, boc-l-pro-nh2, n-tboc-l-prolinamide, r-tert-butyl 2-carbamoylpyrrolidine-1-carboxylate, n-boc-l-prolinamide
PubChem CID 7019053
IUPAC Name tert-butyl (2S)-2-carbamoylpyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)N

Specifications

Melting Point 107°C
Color White
Quantity 1 g
Formula Weight 214.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-prolinamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.