Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-L-proline Methyl Ester 97.0+%, TCI America™
SDP

Catalog No. B522825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B522825G 25 g
B52285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B522825G Supplier TCI America Supplier No. B522825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 59936-29-7
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD01075092
InChI Key WVDGSSCWFMSRHN-QMMMGPOBSA-N
Synonym boc-l-pro-ome, boc-pro-ome, boc-l-proline-methyl ester, s-1-tert-butyl 2-methyl pyrrolidine-1,2-dicarboxylate, boc-l-proline methyl ester, 1-tert-butyl 2-methyl 2s-pyrrolidine-1,2-dicarboxylate, 1-tert-butyl 2-methyl s-pyrrolidine-1,2-dicarboxylate, pubchem18390, boc-l-prolinemethylester, boc-l-prolinem ethylester
PubChem CID 1277666
IUPAC Name 1-tert-butyl 2-methyl (2S)-pyrrolidine-1,2-dicarboxylate
SMILES COC(=O)[C@@H]1CCCN1C(=O)OC(C)(C)C

Specifications

Color Yellow
Boiling Point 135°C
Quantity 25 g
Formula Weight 229.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-proline Methyl Ester
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.