Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-L-tyrosine 98.0+%, TCI America™
SDP

Catalog No. B164010G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B164010G 10 g
B16405G 5 g
B164025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B164010G Supplier TCI America Supplier No. B164010G

Chemical Identifiers

CAS 3978-80-1
Molecular Formula C14H19NO5
Molecular Weight (g/mol) 281.31
MDL Number MFCD00037179
InChI Key CNBUSIJNWNXLQQ-LDGXTIHJNA-N
Synonym boc-tyr-oh, boc-l-tyrosine, n-boc-l-tyrosine, n-tert-butoxycarbonyl-l-tyrosine, n-boc-tyrosine, n-t-boc-l-tyrosine, l-tyrosine, n-1,1-dimethylethoxy carbonyl, chembl65386, n-t-butoxycarbonyl-l-tyrosine, n-tert-butoxycarbonyl tyrosine
PubChem CID 117439
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Melting Point 135°C
Color White
Quantity 10 g
Formula Weight 281.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-tyrosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.