Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-N'-methylethylenediamine 98.0+%, TCI America™
SDP

Catalog No. B40461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40461G 1 g
B40465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40461G Supplier TCI America Supplier No. B40461G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122734-32-1
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.244
MDL Number MFCD06808585
InChI Key GKWGBMHXVRSFRT-UHFFFAOYSA-N
PubChem CID 3435744
IUPAC Name tert-butyl N-[2-(methylamino)ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCCNC

Specifications

Color White-Yellow
Synonym N-(tert-Butoxycarbonyl)-N′C-methyl-1,2-diaminoethane, N-(2-Methylaminoethyl)carbamic Acid tert-Butyl Ester, N-Boc-N′C-methyl-1,2-diaminoethane, N-Boc-N′C-methylethylenediamine, tert-Butyl N-(2-Methylaminoethyl)carbamate
Quantity 1 g
Formula Weight 174.24
Percent Purity ≥98.0% (T)
Physical Form Crystal/Lump
Chemical Name or Material N-(tert-Butoxycarbonyl)-N′-methylethylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.