Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-(tert-Butoxycarbonyl)-trans-4-(p-toluenesulfonyloxy)-L-proline Methyl Ester 98.0+%, TCI America™
SDP

Catalog No. B52471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52471G 1 g
B5247200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52471G Supplier TCI America Supplier No. B52471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88043-21-4
Molecular Formula C18H25NO7S
Molecular Weight (g/mol) 399.458
MDL Number MFCD02179383
InChI Key UKVKNGDXVREPDE-HIFRSBDPSA-N
Synonym boc-trans-4-tosyloxy-l-proline methyl ester, n-boc-trans-4-tosyloxy-l-proline methyl ester, 4r-4-tso-n-boc-pro-ome, n-boc-trans-4-p-tosyloxy-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-tosyloxy pyrrolidine-1,2-dicarboxylate, 2s,4r-4-4-methylphenyl sulfonyl oxy-1,2-pyrrolidinedicarboxylic acid 1-1,1-dimethylethyl 2-methyl ester, 1-tert-butyl 2-methyl 2s,4r-4-4-methylbenzenesulfonyl oxy pyrrolidine-1,2-dicarboxylate, ksc659e9j, boc-trans-4-tosyloxy-l-prolinemethylester
PubChem CID 13425772
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S,4R)-4-(4-methylphenyl)sulfonyloxypyrrolidine-1,2-dicarboxylate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2CC(N(C2)C(=O)OC(C)(C)C)C(=O)OC

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 1 g
Formula Weight 399.46
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-trans-4-(p-toluenesulfonyloxy)-L-proline Methyl Ester
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.