Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-tert-Butylacrylamide 98.0+%, TCI America™
SDP

Catalog No. B0703100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0703100G 100 g
B070325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0703100G Supplier TCI America Supplier No. B0703100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 107-58-4
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.19
MDL Number MFCD00026271
InChI Key XFHJDMUEHUHAJW-UHFFFAOYSA-N
Synonym n-tert-butylacrylamide, tert-butylacrylamide, n-tert-butyl acrylamide, 2-propenamide, n-1,1-dimethylethyl, acrylamide, n-tert-butyl, n-t-butylacrylamide, unii-xj13fsh48k, n-1,1-dimethylethyl-2-propenamide, xj13fsh48k, n-tert-butyl prop-2-enamide
PubChem CID 7877
IUPAC Name N-tert-butylprop-2-enamide
SMILES CC(C)(C)NC(=O)C=C

Specifications

Melting Point 130°C
Color White
Quantity 100 g
Formula Weight 127.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-tert-Butylacrylamide
EINECSNumber (2)-1015
RTECSNumber AS3460000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.