Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Tridecane, 99+%

Catalog No. AC139511000
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC139511000 100 mL Glass bottle
AC139510250 25 mL Glass bottle
AC139515000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC139511000 Supplier Thermo Scientific Chemicals Supplier No. 139511000
Only null left
Add to Cart

CAS: 629-50-5 | C13H28 | 184.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-50-5
Molecular Formula C13H28
Molecular Weight (g/mol) 184.37
MDL Number MFCD00008979
InChI Key IIYFAKIEWZDVMP-UHFFFAOYSA-N
Synonym n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group
PubChem CID 12388
ChEBI CHEBI:35998
IUPAC Name tridecane
SMILES CCCCCCCCCCCCC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -5°C to 4°C
Color Colorless
Density 0.7500g/mL
Boiling Point 234.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4240 to 1.4260
Linear Formula CH3(CH2)11CH3
Quantity 100 mL
Beilstein 01, 171
Specific Gravity 0.75
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol
Formula Weight 184.36
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Tridecane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
EINECSNumber 211-093-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.