Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(Trimethylsilyl)methyl]benzylamine, 95%

Catalog No. AAH6445006
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6445006 5 g
AAH6445022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6445006 Supplier Thermo Scientific Chemicals Supplier No. H6445006

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

CAS: 53215-95-5 | C11H19NSi | 193.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53215-95-5
Molecular Formula C11H19NSi
Molecular Weight (g/mol) 193.37
MDL Number MFCD00012192
InChI Key WECLUYCAWLJMKM-UHFFFAOYSA-N
Synonym n-trimethylsilylmethyl benzylamine, n-trimethylsilyl methyl benzylamine, benzyl trimethylsilyl methyl amine, n-benzyl-1-trimethylsilyl methylamine, 1-phenyl-n-trimethylsilylmethyl methanamine, benzyl trimethylsilylmethyl amine, n-benzyl-1-trimethylsilyl methanamine, benzenemethanamine, n-trimethylsilyl methyl, n-benzylaminomethyltrimethylsilane, n-benzyl-n-trimethylsilyl methyl amine
PubChem CID 353458
SMILES C[Si](C)(C)CNCC1=CC=CC=C1

Specifications

Color Yellow
Density 0.88 g/mL
Boiling Point 89°C to 90°C (5 mmHg)
Flash Point 96°C (205°F)
Refractive Index 1.496
Quantity 5 g
Beilstein 2802488
IUPAC Name 1-phenyl-N-(trimethylsilylmethyl)methanamine
Formula Weight 193.36
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material N-[(Trimethylsilyl)methyl]benzylamine
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.