Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[(Trimethylsilyl)methyl]benzylamine 96.0+%, TCI America™
SDP

Catalog No. T30005ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30005ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T30005ML Supplier TCI America Supplier No. T30005ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53215-95-5
Molecular Formula C11H19NSi
Molecular Weight (g/mol) 193.37
MDL Number MFCD00012192
InChI Key WECLUYCAWLJMKM-UHFFFAOYSA-N
Synonym n-trimethylsilylmethyl benzylamine, n-trimethylsilyl methyl benzylamine, benzyl trimethylsilyl methyl amine, n-benzyl-1-trimethylsilyl methylamine, 1-phenyl-n-trimethylsilylmethyl methanamine, benzyl trimethylsilylmethyl amine, n-benzyl-1-trimethylsilyl methanamine, benzenemethanamine, n-trimethylsilyl methyl, n-benzylaminomethyltrimethylsilane, n-benzyl-n-trimethylsilyl methyl amine
PubChem CID 353458
IUPAC Name benzyl[(trimethylsilyl)methyl]amine
SMILES C[Si](C)(C)CNCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 71°C
Quantity 5 mL
Formula Weight 193.37
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-[(Trimethylsilyl)methyl]benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.