Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[Tris(hydroxymethyl)methyl]acrylamide, 99%

Catalog No. p-7026697
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2285406 5 g
AAB2285414 25 g
AAB2285422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2285406 Supplier Thermo Scientific Chemicals Supplier No. B2285406
Only null left
Add to Cart

CAS: 13880-05-2 | C7H13NO4 | 175.18 g/mol

N-[Tris(hydroxymethyl)methyl] acrylamide is used to prepare N-(5-hydroxymethyl-2-phenyl-[1,3]dioxan-5-yl)-acrylamide by reaction with benzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-[Tris(hydroxymethyl)methyl] acrylamide is used to prepare N-(5-hydroxymethyl-2-phenyl-[1,3]dioxan-5-yl)-acrylamide by reaction with benzaldehyde.

Solubility
Soluble in water.

Notes
Hygroscopic. Light sensitive. Incompatible with strong oxdizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13880-05-2
Molecular Formula C7H13NO4
Molecular Weight (g/mol) 175.18
MDL Number MFCD00081045
InChI Key MVBJSQCJPSRKSW-UHFFFAOYSA-N
Synonym n-tris hydroxymethyl methyl acrylamide, n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl acrylamide, tris-acrylamide, n-acryloyltris hydroxymethyl aminomethane, n-acryloyl-tris hydroxymethyl aminomethane, n-1,3-dihydroxy-2-hydroxymethyl propan-2-yl prop-2-enamide, zlchem 121, acryloylaminomethylidynetrimethanol, n-1,3-dihydroxy-2-hydroxymethyl-2-propanyl acrylamide
PubChem CID 83788
IUPAC Name N-[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]prop-2-enamide
SMILES OCC(CO)(CO)NC(=O)C=C

Specifications

Melting Point 134°C to 138°C
Linear Formula CH2=CHCONHC(CH2OH)3
Quantity 5 g
Sensitivity Hygroscopic; Light sensitive
Solubility Information Soluble in water.
Formula Weight 175.18
Percent Purity 99%
Chemical Name or Material N-[Tris(hydroxymethyl)methyl]acrylamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 237-645-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.