Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Trityl-L-alanine diethylammonium salt, 97%, Thermo Scientific™

Catalog No. AAH6692206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6692206 5 g
AAH6692214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6692206 Supplier Thermo Scientific Chemicals Supplier No. H6692206

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80514-65-4
Molecular Formula C26H32N2O2
Molecular Weight (g/mol) 404.554
MDL Number MFCD02094387
InChI Key PWGPLCPGMCXDCL-LMOVPXPDSA-N
Synonym diethylamine s-2-tritylamino propanoate, trt-ala-oh, 2s-2-triphenylmethyl amino propanoic acid; diethylamine, trt-ala-oh dea, n-alpha-trityl-l-alanine diethylamine salt, n-triphenylmethyl-l-alanine-n-ethylethanamine 1/1
PubChem CID 14283975
IUPAC Name N-ethylethanamine;(2S)-2-(tritylamino)propanoic acid
SMILES CCNCC.CC(C(=O)O)NC(C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Beilstein 3923313
Quantity 5 g
Formula Weight 404.55
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Trityl-L-alanine diethylammonium salt
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.