Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Undecane, 99.7%, MP Biomedicals™

Catalog No. ICN15823180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15823180 100 mL
ICN15823190 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15823180 Supplier MP Biomedicals, Inc Supplier No. 0215823180
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1120-21-4
Molecular Formula C11H24
Molecular Weight (g/mol) 156.31
MDL Number MFCD00008959
InChI Key RSJKGSCJYJTIGS-UHFFFAOYSA-N
Synonym n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und
PubChem CID 14257
ChEBI CHEBI:46342
IUPAC Name undecane
SMILES CCCCCCCCCCC

Specifications

Color Colorless
Density 0.744 at 25°C (water = 1)
Assay Percent Range 0.997
Quantity 100 mL
Formula Weight 156.3
Physical Form Liquid
Chemical Name or Material n-Undecane
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.