Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Undecane, 99%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
10L
1120-21-4
C11H24
156.313
MFCD00008959
RSJKGSCJYJTIGS-UHFFFAOYSA-N
n-undecane,hendecane,n-hendecane,decane, methyl,unii-jv0qt00nue,ccris 3796,jv0qt00nue,undecane,undecane, analytical standard,und
14257
CHEBI:46342
undecane
CCCCCCCCCCC
1120-21-4
C11H24
156.313
MFCD00008959
RSJKGSCJYJTIGS-UHFFFAOYSA-N
n-undecane,hendecane,n-hendecane,decane, methyl,unii-jv0qt00nue,ccris 3796,jv0qt00nue,undecane,undecane, analytical standard,und
14257
CHEBI:46342
undecane
CCCCCCCCCCC
Melting Point -26.0°C
Flash Point 65°C
Color Colorless
Infrared Spectrum Authentic
Boiling Point 196.0°C
Specific Gravity 0.74
Refractive Index 1.4160 to 1.4180
Quantity 10L
Density 0.7400g/mL
Vapor Pressure 0.6hPa at 20°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3(CH2)9CH3
Beilstein 01, IV, 487
Formula Weight 156.31
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material n-Undecane, 99%
Show More Show Less

GHS H Statement:
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 214-300-6

RTECSNumber : YQ1525000

TSCA : TSCA