Learn More
IPHASE PHARMA SERVICES LLC New Zealand White Rabbit Liver Microsomes,Male,0.5mL,20mg/mL
Supplier: IPHASE PHARMA SERVICES LLC 0121I1.01
A major tool for metabolism stability study. Our liver microsomes products available in multi-species (human, monkey, beagle, rat, and mouse). We also offer customized products prepared from unconventional animal species, disease models, or animals of certain ages.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.