Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ Naltrexone Solution, 1.0 mg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco N0071ML

Catalog No. 01-001-703


Only null left
Add to Cart

Description

Description

Naltrexone is an opiate sold under the trade names Revia, Depade and Vivitrol™ and is prescribed primarily for the management of alcohol or opiate dependence. This analytical reference standard is applicable for use in LC/MS or GC/MS applications such as pharmaceutical research, forensic analysis, clinical toxicology, pain prescription monitoring or urine drug testing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1, 16590-41-3
MFCD00242996
1.0 mg/mL in methanol
Ampule
9.7°C (49.5°F) - closed cup
C20H23NO4
UN 1230 - class 3 - PG 2 - Methanol, solution
341.40
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

Recommended Storage : 2 to 8°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

CERILLIANT is a registered trademark of Cerilliant Corporation