Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CERILLIANT™ (+/-)-Nornicotine-d4, 100 μg/mL in Methanol, Sold by MilliporeSigma™ Supelco™

Certified reference material

Supplier:  MilliporeSigma Supelco N0721ML

Catalog No. 01-001-740


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

An internal standard for nicotine quantitation by LC/MS or GC/MS. Nornicotine, a tobacco alkaloid and nicotine metabolite, is used as an indicator of smoking and tobacco use or nicotine replacement products. This Certified Spiking Solution™ is suitable for use in nicotine testing by LC-MS/MS or GC/MS.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

67-56-1, 66148-18-3
UN 1230 - class 3 - PG 2 - Methanol, solution
(+\-)-Nornicotine-d4 solution
Ampule
9.7°C (49.5°F) - closed cup
C13H20N2
100 μg/mL in methanol
204.31
1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

H331-H370

H311

EINECSNumber : 200-659-6

Recommended Storage : -20°C

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

CERILLIANT is a registered trademark of Cerilliant Corporation