Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N1-Acetylsulfamethoxazole, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013392 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3392 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3392 Supplier MilliporeSigma Supelco Supplier No. 9159425MG

Specifications

CAS 18607-98-2
Linear Formula C12H13N3O4 S
Synonym N-[(4-Aminophenyl)sulfonyl]-N-(5-methyl-3-isoxazolyl)acetamide; N1-Acetyl-N1-(5-methyl-3-isoxazolyl)sulfanilamide
Type Analytical Standard
Quantity 25 mg
Formula Weight 295.31
Percent Purity ≥95% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.