Learn More
Apexbio Technology LLC N1-Methyl-2'-O-Methylpseudo-UTP(Synonyms: N1-Methyl-2'-O-Methylpseudo-UTP, N1-Me-2'-O-Me-pseudo-UTP, 1-Methylpseudouridine-5'-triphosphate-2'-O-methyl), 10ul (100 mM).
SDP
Supplier:  Apexbio Technology LLC B80700.01
Encompass_Preferred
N1-Methyl-2'-O-Methylpseudo-UTP is a chemically modified uridine triphosphate analog, incorporating an N1-methylpseudouridine base and a 2'-O-methyl ribose modification. Structurally designed to substitute natural UTP during in vitro RNA synthesis, this compound is efficiently incorporated into RNA transcripts by T7 RNA polymerase. RNAs synthesized using this modified nucleotide exhibit reduced immunogenic responses, facilitating investigations into RNA structure-function relationships, innate immune sensing mechanisms, and the development of messenger RNA therapeutics.
Catalog No. 50-000-60675
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.