Learn More
FUJIFILM BIOSCIENCES INC N2 Supplement with Transferrin
Supplier:  FUJIFILM BIOSCIENCES INC 14109041
Encompass_Preferred
This product is a general-purpose serum substitute used for the culture of nerve cells It is suited to primary cultures of nerve cells and nerve stem cells As differentiation of nerve stem cells is induced by the components contained in FBS a serum substitute such as this product is used to culture nerve stem cells in undifferentiated condition Transferrin apo is used in this product It has been confirmed that some cell types proliferate favorably in N2 supplement using transferrin apo than that using transferrin holo
Catalog No. 50-004-50436