Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ N21-MAX Vitamin A Free Media Supplement (50X)
SDP

Catalog No. AR012 Shop All Novus Biologicals Products
Change view
Click to view available options
N21-MAX Vitamin A free media supplement; 50X; Qty: 10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AR012 N21-MAX Vitamin A free media supplement; 50X; Qty: 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
This product contains human transferrin. This transferrin was tested at the donor level using an FDA licensed method and found to be non-reactive for anti-HIV-1/2 and Hepatitis B surface antigen. As no testing can offer complete assurance of freedom from infectious agents, these reagents should be handled as if capable of transmitting infection.
Catalog No. AR012 Supplier R&D Systems™ Supplier No. AR012

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This product contains human transferrin. This transferrin was tested at the donor level using an FDA licensed method and found to be non-reactive for anti-HIV-1/2 and Hepatitis B surface antigen. As no testing can offer complete assurance of freedom from infectious agents, these reagents should be handled as if capable of transmitting infection.

Used for controlled neural stem and progenitor cell expansion. R&D Systems N21-MAX Vitamin A Free Media Supplement (50X) is a nutrient-rich, serum-free, and fully defined media formulation.

  • Supplied as 50X concentrated solution
  • Limits retinoic acid production to reduce unwanted NSC differentiation
  • Rigorous quality control testing ensures consistency across experiments
  • Eliminates the need for an astrocyte feeder layer
  • Used to mature and maintain healthy primary neurons in culture
  • Quality tested to reduce unwanted experimental variation
  • Contains factors qualified to the expansion of NSCs in vitro
  • Does not require an astrocyte feeder layer

Specifications

Concentration 50X
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Form Liquid
Product Type Media Supplement
Includes Albumin (bovine), L-Carnitine, Catalase, Corticosterone, Ethanolamine, Glutathione, Galactose, Holo-Transferrin, Insulin, Linoleic Acid, Linolenic Acid, Lipoic Acid, Progesterone, Putrescine, Selenite, Superoxide dismutase, Triiodo-L-thyronine, D,L-alpha-Tocopherol, D,L-alpha-Tocopherol acetate. Supplied in a volume sufficient to supplement 500mL of media at the recommended concentration. N21-MAX Vitamin A Free Media Supplement is tested for use in Neurobasal Media.
Quantity 10 mL
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.