Learn More
Sigma Aldrich Fine Chemicals Biosciences N3 PEG5K NHS avg Mn 50500MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences JKA5088500MG
Encompass
Azide PEG NHS Ester. The Azide group may be reduced to amine by hydrogenolysis Click chemistry PEG reagent for reaction with alkynes.
Catalog No. 50-290-7290
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.